1,4-Diaminobutane dihydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18312-14
A18312-22
CAAAA18312-14EA
122.77
CAD
CAAAA18312-14
CAAAA18312-22
1,4-Diaminobutane dihydrochloride 99%
1,4-Diaminobutane dihydrochloride
Purity: 99%
CAS No.: 333-93-7
Molecular Formula: HCl.H2N(CH2)4NH2.HCl
Formula Weight: 161.08
Melting Point: ca 280° dec.
Beilstein Reference: 3906680
Merck Reference: 14,7947
MDL No.: MFCD00012526
Formula:
C₄H₁₄Cl₂N₂ Melting Pt: ca 280 °C (dec.) Density: 1.31 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012526 CAS Number: 333-93-7 EINECS: 206-375-9 Merck Index: 14,7947 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...