N,N-Dimethyl-4-nitrosoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18301-30
A18301-14
CAAAAA18301-30EA
371.3
CAD
CAAAAA18301-30
CAAAA18301-14
N,N-Dimethyl-4-nitrosoaniline 98%
N,N-Dimethyl-4-nitrosoaniline
Purity: 98%
CAS No.: 138-89-6
Molecular Formula: C8H10N2O
Formula Weight: 150.18
Melting Point: 84-87°
Beilstein Reference: 607293
Merck Reference: 14,6638
UN No.: 1369
MDL No.: MFCD00002063
Formula:
C₈H₁₀N₂O Boiling Pt: 259 °C (1013 hPa) Melting Pt: 84…87 °C Density: 1.150 Storage Temperature: Ambient |
MDL Number:
MFCD00002063 CAS Number: 138-89-6 EINECS: 205-343-1 UN: 1369 ADR: 4.2,II Merck Index: 14,6638 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...