1-Butanesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Butanesulphonylchloride, 1-Butanesulfonylchloride

A18295-22 A18295-06
CAAAAA18295-22EA 404.48 CAD
CAAAAA18295-22 CAAAA18295-06
1-Butanesulfonyl chloride 98%
1-Butanesulphonyl chloride

Purity: 98% CAS No.: 2386-60-9 Molecular Formula: C4H9ClO2S Formula Weight: 156.63 Flash Point: 79°(174°F) Refractive Index: 1.4560 Melting Point: -29° Boiling Point: 80-81°/9mm Beilstein Reference: 1748742 UN No.: 3265 MDL No.: MFCD00007464

Formula: C₄H₉ClO₂S
Boiling Pt: 80…81 °C/9 mm
Melting Pt: –29 °C
Density: 1.210
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007464
CAS Number: 2386-60-9
EINECS: 219-204-8
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR