2,3,5,6-Tetramethylpyrazine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18279-14
A18279-22
CAAAA18279-14EA
80.42
CAD
CAAAA18279-14
CAAAA18279-22
2,3,5,6-Tetramethylpyrazine 98+%
2,3,5,6-Tetramethylpyrazine
Purity: 98+%
CAS No.: 1124-11-4
Molecular Formula: C8H12N2
Formula Weight: 136.20
Odor: Pleasant
pH: 7
Melting Point: 83-86°
Boiling Point: 190°
Beilstein Reference: 113100
MDL No.: MFCD00006146
Formula:
C₈H₁₂N₂ MW: 136.2 g/mol Boiling Pt: 190 °C Melting Pt: 83…86 °C Density: 1.08 Storage Temperature: Ambient |
MDL Number:
MFCD00006146 CAS Number: 1124-11-4 EINECS: 214-391-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...