Phenyl propargyl ether 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18272-03
A18272-06
CAAAA18272-03EA
64.46
CAD
CAAAA18272-03
CAAAA18272-06
Phenyl propargyl ether 98+%
Phenyl propargyl ether
Purity: 98+%
CAS No.: 13610-02-1
Molecular Formula: C9H8O
Formula Weight: 132.16
Storage Temperature: 2°C to 8°C
Flash Point: 76°(169°F)
Refractive Index: 1.5330
Odor: Pungent
Boiling Point: 89-90°/14mm
MDL No.: MFCD00048107
Formula:
C₉H₈O Boiling Pt: 89…90 °C/14 mm Density: 1.030 Flash Pt: 76 °C(169 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00048107 CAS Number: 13610-02-1 EINECS: 237-095-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...