4-Nitroveratrole 98+%

Supplier: Thermo Scientific Chemicals
Warning

A18264-18 A18264-30 A18264-14
CAAAAA18264-18EA 110.76 CAD
CAAAAA18264-18 CAAAAA18264-30 CAAAA18264-14
4-Nitroveratrole 98+%
4-Nitroveratrole

Purity: 98+% CAS No.: 709-09-1 Molecular Formula: C8H9NO4 Formula Weight: 183.16 Melting Point: 95-98° Boiling Point: 221-223°/10mm MDL No.: MFCD00007238

Formula: C₈H₉NO₄
Boiling Pt: 221…223 °C (10 mmHg)
Melting Pt: 95…98 °C
Density: 1.1888 g/cm³ (132.7 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007238
CAS Number: 709-09-1
EINECS: 211-906-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR