meso-Tetraphenylporphine low chlorine

Supplier: Thermo Scientific Chemicals
Warning

A18261-03 A18261-06 A18261-14
CAAAA18261-03EA 105.39 CAD
CAAAA18261-03 CAAAA18261-06 CAAAA18261-14
meso-Tetraphenylporphine low chlorine
meso-Tetraphenylporphine

CAS No.: 917-23-7 Molecular Formula: C44H30N4 Formula Weight: 614.71 Melting Point: 449-451° Beilstein Reference: 379542 MDL No.: MFCD00011680

Formula: C₄₄H₃₀N₄
Melting Pt: 449…451 °C
Density: 1.276 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011680
CAS Number: 917-23-7
EINECS: 213-025-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR