1-Chloro-2-iodobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-1-iodobenzene

A18255-09 A18255-14 A18255-22
CAAAAA18255-09EA 57.2 CAD
CAAAAA18255-09 CAAAA18255-14 CAAAA18255-22
1-Chloro-2-iodobenzene 99%
1-Chloro-2-iodobenzene

Purity: 99% CAS No.: 615-41-8 Molecular Formula: C6H4ClI Formula Weight: 238.46 Flash Point: 111°(231°F) Refractive Index: 1.6345 Melting Point: 1° Boiling Point: 108-110°/15mm Beilstein Reference: 1905116 MDL No.: MFCD00001033

Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 108…110 °C (15 mmHg)
Melting Pt: 1 °C
Density: 1.954
Flash Pt: 111 °C (231 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001033
CAS Number: 615-41-8
EINECS: 210-423-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR