2-Chlorophenyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Chloro-2-isothiocyanatobenzene

A18238-06 A18238-14
CAAAA18238-06EA 69.84 CAD
CAAAA18238-06 CAAAA18238-14
2-Chlorophenyl isothiocyanate 97%
2-Chlorophenyl isothiocyanate

Purity: 97% CAS No.: 2740-81-0 Molecular Formula: C7H4ClNS Formula Weight: 169.64 Flash Point: 110°(230°F) Refractive Index: 1.6608 Odor: Pungent Boiling Point: 260-262° Beilstein Reference: 2081473 UN No.: 2922 MDL No.: MFCD00004801

Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 260…262 °C
Density: 1.287
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004801
CAS Number: 2740-81-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR