Dibutyl sebacate 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18231-22
A18231-36
CAAAA18231-22EA
56.72
CAD
CAAAA18231-22
CAAAA18231-36
Dibutyl sebacate 90+%
Dibutyl sebacate
Purity: 90+%
CAS No.: 109-43-3
Molecular Formula: CH3(CH2)3O2C(CH2)8CO2(CH2)3CH3
Formula Weight: 314.47
Flash Point: 178°(352°F)
Refractive Index: 1.441
Melting Point: -11°
Boiling Point: 344-345°
Beilstein Reference: 1798308
MDL No.: MFCD00027218
Formula:
C₁₈H₃₄O₄ Boiling Pt: 344…345 °C Melting Pt: –11 °C Density: 0.94 Flash Pt: 178 °C(352 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027218 CAS Number: 109-43-3 EINECS: 203-672-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...