3-Chloro-o-toluidine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Amino-6-chlorotoluene, 3-Chloro-2-methylaniline
A18221-0B
A18221-30
CAAAA18221-0BEA
679.4
CAD
CAAAA18221-0B
CAAAA18221-30
3-Chloro-o-toluidine 98+%
3-Chloro-o-toluidine
Purity: 98+%
CAS No.: 87-60-5
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 113°(235°F)
Refractive Index: 1.5890
Melting Point: 2°
Boiling Point: 245°
Beilstein Reference: 471697
UN No.: 3429
MDL No.: MFCD00007766
Formula:
C₇H₈ClN MW: 141.6 g/mol Boiling Pt: 245 °C Melting Pt: 2 °C Density: 1.188 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007766 CAS Number: 87-60-5 EINECS: 201-756-6 UN: 3429 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...