Nonylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18209-14
A18209-22
A18209-36
CAAAA18209-14EA
186.44
CAD
CAAAA18209-14
CAAAA18209-22
CAAAA18209-36
Nonylamine 97%
Nonylamine
Purity: 97+%
CAS No.: 112-20-9
Molecular Formula: CH3(CH2)8NH2
Formula Weight: 143.27
Flash Point: 62°(143°F)
Refractive Index: 1.4330
Odor: Amine-like
Melting Point: -1°
Boiling Point: 200-202°
Beilstein Reference: 1733633
UN No.: 2735
MDL No.: MFCD00008249
Formula:
H₃C(CH₂)₇CH₂NH₂ Boiling Pt: 200…202 °C Melting Pt: –1 °C Density: 0.784 Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008249 CAS Number: 112-20-9 EINECS: 203-945-9 UN: 2735 ADR: 8,III |
Specification Test Results
Refractive index | 1.4330 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...