α-Bromo-4-iodotoulene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18200-06
A18200-14
CAAAA18200-06EA
144.57
CAD
CAAAA18200-06
CAAAA18200-14
CAAAA18200-22
α-Bromo-4-iodotoulene 97%
α-Bromo-4-iodotoulene
Purity: 97%
CAS No.: 16004-15-2
Molecular Formula: C7H6BrI
Formula Weight: 296.94
Odor: Irritating
Melting Point: 78-82°
Boiling Point: 140°/15mm
Beilstein Reference: 2325160
UN No.: 3261
MDL No.: MFCD00209656
Formula:
C₇H₆BrI Boiling Pt: 140 °C/15 mm Melting Pt: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00209656 CAS Number: 16004-15-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...