3-Cyclopentylpropionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A18192-14 A18192-22
CAAAA18192-14EA 92.75 CAD
CAAAA18192-14 CAAAA18192-22
3-Cyclopentylpropionic acid 98%
3-Cyclopentylpropionic acid

Purity: 98% CAS No.: 140-77-2 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 47°(117°F) Refractive Index: 1.4570 Boiling Point: 130-133°/12mm UN No.: 1993 MDL No.: MFCD00001392

Formula: C₈H₁₄O₂
Boiling Pt: 130…133 °C/12 mm
Density: 0.996
Flash Pt: 47 °C (117 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001392
CAS Number: 140-77-2
EINECS: 205-433-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR