3-Aminophenylboronic acid monohydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dihydroxy(3-aminophenyl)borane monohydrate, 3-Aminobenzeneboronic acid monohydrate, 3-Aminobenzeneboronic acid, 3-Aminophenylboronic Acid Monohydrate, (3-Aminophenyl)metaboric acid monohydrate

A18189-03 A18189-06
CAAAA18189-03EA 95.12 CAD
CAAAA18189-03 CAAAA18189-06
3-Aminophenylboronic acid monohydrate 97%
3-Aminophenylboronic acid monohydrate

Purity: 97% CAS No.: 206658-89-1 Molecular Formula: C6H8BNO2.H2O Formula Weight: 154.96 (136.95anhy) Melting Point: 93-97° Beilstein Reference: 2936342

Formula: C₆H₈BNO₂·1H₂O
MW: 154.96 g/mol
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00149554
CAS Number: 206658-89-1
EINECS: 250-189-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR