3,5-Heptanedione 98%

Supplier: Thermo Scientific Chemicals
Warning

A18165-06 A18165-14
CAAAA18165-06EA 102.07 CAD
CAAAA18165-06 CAAAA18165-14
3,5-Heptanedione 98%
3,5-Heptanedione

Purity: 98% CAS No.: 7424-54-6 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 57°(134°F) Refractive Index: 1.4555 Odor: Pleasant Boiling Point: 175-177° Beilstein Reference: 635979 UN No.: 1224 MDL No.: MFCD00015186

Formula: C₇H₁₂O₂
Boiling Pt: 175…177 °C
Density: 0.8
Flash Pt: 57 °C (134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015186
CAS Number: 7424-54-6
EINECS: 231-054-5
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR