N,N-Diethyl-1,1,1-trimethylsilylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18149-14
A18149-22
CAAAA18149-14EA
84.85
CAD
CAAAA18149-14
CAAAA18149-22
N,N-Diethyl-1,1,1-trimethylsilylamine 98%
N,N-Diethyl-1,1,1-trimethylsilylamine
Purity: 98%
CAS No.: 996-50-9
Molecular Formula: (CH3)3SiN(CH2CH3)2
Formula Weight: 145.32
Flash Point: 10°(50°F)
Refractive Index: 1.4110
Odor: Amine-like
Melting Point: -10°
Boiling Point: 125-126°
Beilstein Reference: 635718
UN No.: 2733
MDL No.: MFCD00009040
Formula:
C₇H₁₉NSi Boiling Pt: 125…126 °C Melting Pt: –10 °C Density: 0.760 Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009040 CAS Number: 996-50-9 EINECS: 213-637-6 UN: 2733 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...