1,4-Dibromotetrafluorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

A18135-03 A18135-06 A18135-14
CAAAA18135-03EA 62.25 CAD
CAAAA18135-03 CAAAA18135-06 CAAAA18135-14
1,4-Dibromotetrafluorobenzene 99%
1,4-Dibromotetrafluorobenzene

Purity: 99% CAS No.: 344-03-6 Molecular Formula: C6Br2F4 Formula Weight: 307.87 Melting Point: 78-81° MDL No.: MFCD00000309

Formula: C₆Br₂F₄
MW: 307.87 g/mol
Boiling Pt: 198
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00000309
CAS Number: 344-03-6
EINECS: 206-448-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR