2-Fluoro-5-nitrobenzotrifluoride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-5-nitrobenzotrifluoride, 1-Fluoro-4-nitro-2-(trifluoromethyl)benzene
A18130-06
A18130-14
CAAAA18130-06EA
52.3
CAD
CAAAA18130-06
CAAAA18130-14
2-Fluoro-5-nitrobenzotrifluoride 98%
2-Fluoro-5-nitrobenzotrifluoride
Purity: 98%
CAS No.: 400-74-8
Molecular Formula: C7H3F4NO2
Formula Weight: 209.10
Flash Point: 91°(195°F)
Refractive Index: 1.4650
Boiling Point: 105-110°/25mm
Beilstein Reference: 1881423
UN No.: 2810
MDL No.: MFCD00024576
Formula:
C₇H₃F₄NO₂ MW: 209.1 g/mol Boiling Pt: 105…110 °C (25 mmHg) Melting Pt: 22…24 °C Density: 1.522 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00024576 CAS Number: 400-74-8 EINECS: 206-925-8 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...