Tribromofluoromethane 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
A18124-14
A18124-18
A18124-06
CAAAAA18124-14EA
481.9
CAD
CAAAAA18124-14
CAAAAA18124-18
CAAAA18124-06
Tribromofluoromethane 99% stabilized
Tribromofluoromethane
Purity: 99%
CAS No.: 353-54-8
Molecular Formula: CBr3F
Formula Weight: 270.72
Flash Point: None
Refractive Index: 1.5255
Melting Point: -74°
Boiling Point: 106-107°
Beilstein Reference: 1732796
MDL No.: MFCD00000366
Formula:
Br₃CF Boiling Pt: 106…107 °C Melting Pt: –74 °C Density: 2.765 Storage Temperature: Ambient |
MDL Number:
MFCD00000366 CAS Number: 353-54-8 EINECS: 206-535-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...