Isopropyl myristate 98%
Supplier: Thermo Scientific Chemicals
A18111-0F
A18111-AE
A18111-AP
CAAAA18111-0FEA
606.72
CAD
CAAAA18111-0F
CAAAA18111-AE
CAAAA18111-AP
Isopropyl myristate 98%
Isopropyl myristate
Purity: 98%
CAS No.: 110-27-0
Molecular Formula: CH3(CH2)12CO2CH(CH3)2
Formula Weight: 270.46
Flash Point: 152°(306°F)
Physical Form: Liquid
Density: 0.85
Appearance: Colorless
Boiling Point: 191-194°/20mm
MDL No.: MFCD00008982
Formula:
C₁₇H₃₄O₂ Boiling Pt: 191…194 °C/20 mm Melting Pt: 0…1 °C Density: 0.85 Flash Pt: 152 °C(306 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008982 CAS Number: 110-27-0 EINECS: 203-751-4 Merck Index: 13,05235 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...