3-Hydroxyflavone 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 3-Hydroxy-2-phenyl-4H-chromen-4-one, 3-Hydroxy-2-phenyl-4-benzopyrone, 3-Hydroxy-2-phenylchromen-4-one, Flavonol, 3-Hydroxy-2-phenyl-4H-1-benzopyran-4-one

A18110-03 A18110-06
CAAAA18110-03EA 55.3 CAD
CAAAA18110-03 CAAAA18110-06
3-Hydroxyflavone 98+%
3-Hydroxyflavone

Purity: 98+% CAS No.: 577-85-5 Molecular Formula: C15H10O3 Formula Weight: 238.25 Melting Point: 168-172° Beilstein Reference: 15789

Formula: C₁₅H₁₀O₃
MW: 238.24 g/mol
Boiling Pt: 411 °C (1013 hPa)
Melting Pt: 168…172 °C
Density: 1.393 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006832
CAS Number: 577-85-5
EINECS: 209-416-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR