Isonicotinic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
A18109-0E
A18109-22
A18109-36
CAAAAA18109-0EEA
486.64
CAD
CAAAAA18109-0E
CAAAA18109-22
CAAAA18109-36
Isonicotinic acid 99%
Isonicotinic acid
Purity: 99%
CAS No.: 55-22-1
Molecular Formula: C6H5NO2
Formula Weight: 123.11
pH: 3.0-4.0
Melting Point: ca 310° subl.
Boiling Point: 260°/15mm
Beilstein Reference: 109599
Merck Reference: 14,5187
MDL No.: MFCD00006429
Formula:
C₆H₅NO₂ MW: 123.11 g/mol Boiling Pt: 260 °C/15 mm Melting Pt: ∼ 310 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00006429 CAS Number: 55-22-1 EINECS: 200-228-2 Merck Index: 14,5187 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...