trans-2-Nonenal 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E)-2-Nonenal

A18108-06 A18108-14
CAAAA18108-06EA 78.53 CAD
CAAAA18108-06 CAAAA18108-14
trans-2-Nonenal 97%
trans-2-Nonenal

Purity: 97% CAS No.: 18829-56-6 Molecular Formula: C9H16O Formula Weight: 140.22 Storage Temperature: 2°C to 8°C Flash Point: 84°(184°F) Refractive Index: 1.4403 Boiling Point: 98-100°/12mm Merck Reference: 14,6676 MDL No.: MFCD00007012

Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 98…100 °C/12 mm
Density: 0.846
Flash Pt: 84 °C(184 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007012
CAS Number: 18829-56-6
EINECS: 242-609-6
Merck Index: 14,6676

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR