Carboxymethyl cellulose sodium salt
Supplier: Thermo Scientific Chemicals
A18105-0E
A18105-36
CAAAA18105-0EEA
423.44
CAD
CAAAA18105-0E
CAAAA18105-36
Carboxymethyl cellulose sodium salt
Carboxymethyl cellulose sodium salt
CAS No.: 9004-32-4
Melting Point:
Formula:
[C₆H₇O₂(OH)x(OCH₂COONa)y]n Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081472 CAS Number: 9004-32-4 Merck Index: 14,1829 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...