4-Nitrobenzenesulfonamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18101-06
A18101-14
CAAAA18101-06EA
86.58
CAD
CAAAA18101-06
CAAAA18101-14
4-Nitrobenzenesulfonamide 97%
4-Nitrobenzenesulphonamide
Purity: 97%
CAS No.: 6325-93-5
Molecular Formula: C6H6N2O4S
Formula Weight: 202.19
Melting Point: 178-181°
MDL No.: MFCD00007937
Formula:
C₆H₆N₂O₄S Melting Pt: 178…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007937 CAS Number: 6325-93-5 EINECS: 228-691-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...