2,6-Difluorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18099-06
A18099-14
CAAAA18099-06EA
92.27
CAD
CAAAA18099-06
CAAAA18099-14
2,6-Difluorobenzaldehyde 97%
2,6-Difluorobenzaldehyde
Purity: 97%
CAS No.: 437-81-0
Molecular Formula: C7H4F2O
Formula Weight: 142.11
Flash Point: 73°(163°F)
Refractive Index: 1.5070
Melting Point: 15-17°
Boiling Point: 85-86°/20mm
Beilstein Reference: 1935273
MDL No.: MFCD00010293
Formula:
C₇H₄F₂O Boiling Pt: 85…86 °C/20 mm Melting Pt: 15…17 °C Density: 1.317 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010293 CAS Number: 437-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...