α,α,α,2-Tetrafluorotoluene 99%

Supplier: Thermo Scientific Chemicals
Danger

A18095-06 A18095-14 A18095-22
CAAAA18095-06EA 46.61 CAD
CAAAA18095-06 CAAAA18095-14 CAAAA18095-22
α,α,α,2-Tetrafluorotoluene 99%
α,α,α,2-Tetrafluorotoluene

Purity: 99% CAS No.: 392-85-8 Molecular Formula: C7H4F4 Formula Weight: 164.10 Flash Point: 17°(62°F) Refractive Index: 1.4060 Melting Point: -49° Boiling Point: 114-115° Beilstein Reference: 2046638 UN No.: 1993 MDL No.: MFCD00000374

Formula: C₇H₄F₄
Boiling Pt: 114…115 °C
Melting Pt: –49 °C
Density: 1.293
Flash Pt: 17 °C (62 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000374
CAS Number: 392-85-8
EINECS: 206-880-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR