2-Methoxyphenyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Isocyanato-2-methoxybenzene

A18075-06 A18075-14
CAAAA18075-06EA 64.15 CAD
CAAAA18075-06 CAAAA18075-14
2-Methoxyphenyl isocyanate 98%
2-Methoxyphenyl isocyanate

Purity: 98% CAS No.: 700-87-8 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5414 Odor: Pungent Boiling Point: 94-96°/10mm Beilstein Reference: 387700 UN No.: 2206 MDL No.: MFCD00002004

Formula: C₈H₇NO₂
Boiling Pt: 94…96 °C/10 mm
Density: 1.122
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002004
CAS Number: 700-87-8
EINECS: 211-853-5
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR