3-Hydroxybenzophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18061-03
A18061-06
A18061-14
CAAAA18061-03EA
89.74
CAD
CAAAA18061-03
CAAAA18061-06
CAAAA18061-14
3-Hydroxybenzophenone 99%
3-Hydroxybenzophenone
Purity: 99%
CAS No.: 13020-57-0
Molecular Formula: C13H10O2
Formula Weight: 198.22
Melting Point: 113-115°
MDL No.: MFCD00002297
Formula:
C₁₃H₁₀O₂ Boiling Pt: 148 °C (4 mmHg) Melting Pt: 113…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002297 CAS Number: 13020-57-0 EINECS: 235-879-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...