2-Bromo-2-butene cis- and trans-mixture 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18058-14
A18058-22
CAAAA18058-14EA
175.38
CAD
CAAAA18058-14
CAAAA18058-22
2-Bromo-2-butene cis- and trans-mixture 98% stabilized
2-Bromo-2-butene
Purity: 98%
CAS No.: 13294-71-8
Molecular Formula: C4H7Br
Formula Weight: 135.00
Storage Temperature: 2°C to 8°C
Flash Point: 1°(33°F)
Refractive Index: 1.4610
Odor: Recognizable
Boiling Point: 82-90°
Beilstein Reference: 1733329
UN No.: 1993
MDL No.: MFCD00000141
Formula:
C₄H₇Br Boiling Pt: 82…90 °C Density: 1.328 Flash Pt: 1 °C (33 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000141 CAS Number: 13294-71-8 EINECS: 236-316-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...