5-Bromovaleryl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18046-06
A18046-14
CAAAA18046-06EA
89.9
CAD
CAAAA18046-06
CAAAA18046-14
5-Bromovaleryl chloride 98%
5-Bromovaleryl chloride
Purity: 98%
CAS No.: 4509-90-4
Molecular Formula: C5H8BrClO
Formula Weight: 199.46
Flash Point: 107°(225°F)
Refractive Index: 1.4910
Odor: Irritating
Boiling Point: 43-45°/0.2mm
Beilstein Reference: 1745663
UN No.: 3265
MDL No.: MFCD00013660
Formula:
BrH₂C(CH₂)₃COOCl Boiling Pt: 43…45 °C/0.2 mm Density: 1.49 Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013660 CAS Number: 4509-90-4 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...