Oleyl alcohol tech. 80-85%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: cis-Octadec-9-en-1-olcis-9-Octadecen-1-ol

A18018-36 A18018-22 A18018-22 A18018-36
CAAAA18018-22EA 67.31 CAD
AAA18018-36 CAAAA18018-22 CAAAA18018-36 AAA18018-22
Oleyl alcohol tech. 80-85%, Technical Grade
Oleyl alcohol
Formula: C₁₈H₃₆O
MW: 268.48 g/mol
Boiling Pt: 206…207 °C/13 mm
Melting Pt: ∼ 6 °C
Density: 0.849
Flash Pt: 180 °C (356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002993
CAS Number: 143-28-2
Merck Index: 14,6831

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR