Dioctylamine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Di-n-octylamine, N-Octyl-1-octanamine
A18017-14
A18017-22
CAAAA18017-14EA
54.83
CAD
CAAAA18017-14
CAAAA18017-22
Dioctylamine 96%
Dioctylamine
Purity: 98%
CAS No.: 1120-48-5
Molecular Formula: C16H35N
Formula Weight: 241.46
Flash Point: 142°(287°F)
Refractive Index: 1.4432
Odor: Amine-like
Melting Point: 13-15°
Boiling Point: 297-298°
Beilstein Reference: 1748376
UN No.: 2735
MDL No.: MFCD00009557
Formula:
C₁₆H₃₅N MW: 241.46 g/mol Boiling Pt: 297…298 °C Melting Pt: 13…15 °C Density: 0.799 Flash Pt: 142 °C (287 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009557 CAS Number: 1120-48-5 EINECS: 214-311-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...