4-Nitrobenzhydrazide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A18016-09 A18016-18
CAAAAA18016-30EA 1249.07 CAD
CAAAAA18016-30 CAAAA18016-09 CAAAA18016-18
4-Nitrobenzhydrazide 98+%
4-Nitrobenzhydrazide

Purity: 98+% CAS No.: 636-97-5 Molecular Formula: C7H7N3O3 Formula Weight: 181.15 Melting Point: ca 216° dec. Beilstein Reference: 519882

Formula: C₇H₇N₃O₃
Melting Pt: ∼ 216 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007604
CAS Number: 636-97-5
EINECS: 211-271-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR