L(-)-Glutathione (reduced form) 97%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

A18014-06 A18014-14 A18014-22
CAAAA18014-06EA 82.16 CAD
CAAAA18014-06 CAAAA18014-14 CAAAA18014-22
L(-)-Glutathione (reduced form) 97%
L(-)-Glutathione (reduced form)

Purity: 97% CAS No.: 70-18-8 Molecular Formula: C10H17N3O6S Formula Weight: 307.33 Storage Temperature: 2°C to 8°C Melting Point: ca 193° dec. Beilstein Reference: 1729812 Merck Reference: 14,4475 Optical Rotation: -17° (c=2 in water) MDL No.: MFCD00065939

Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
Melting Pt: ∼ 193 °C (dec.)
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 14,4475

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR