Oxalyl dichloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18012-22
A18012-0B
A18012-18
A18012-30
CAAAAA18012-22EA
128.14
CAD
CAAAAA18012-22
CAAAA18012-0B
CAAAA18012-18
CAAAA18012-30
Oxalyl dichloride 98%
Oxalyl dichloride
Purity: 98%
CAS No.: 79-37-8
Molecular Formula: C2Cl2O2
Formula Weight: 126.93
Physical Form: Liquid
Density: 1.455
Refractive Index: 1.4290
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Acrid
Melting Point: -12°
Boiling Point: 63-64°
Beilstein Reference: 1361988
Merck Reference: 14,6914
UN No.: 2927
MDL No.: MFCD00000704
Formula:
C₂Cl₂O₂ Boiling Pt: 63…64 °C Melting Pt: –12 °C Density: 1.455 Flash Pt: ≥ 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000704 CAS Number: 79-37-8 EINECS: 201-200-2 UN: 3130 ADR: 4.3,III Merck Index: 14,6914 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...