D-(-)-Diethyl tartrate 99%

Supplier: Thermo Scientific Chemicals
Warning

A17992-06 A17992-14 A17992-22
CAAAA17992-06EA 44.71 CAD
CAAAA17992-06 CAAAA17992-14 CAAAA17992-22
D-(-)-Diethyl tartrate 99%
D(-)-Diethyl tartrate

Purity: 99% CAS No.: 13811-71-7 Molecular Formula: C8H14O6 Formula Weight: 206.19 Flash Point: 93°(199°F) Refractive Index: 1.4460 Melting Point: 17° Boiling Point: 280° Beilstein Reference: 1727143 Optical Rotation: -26.5° (c=1 in water) MDL No.: MFCD00064451

Formula: C₈H₁₄O₆
Boiling Pt: 280 °C
Melting Pt: 17 °C
Density: 1.205
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064451
CAS Number: 13811-71-7
EINECS: 237-458-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR