2-Butoxyethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether

A17976-0D A17976-0F A17976-AP
CAAAA17976-0DEA 372.88 CAD
CAAAA17976-0D CAAAA17976-0F CAAAA17976-AP
2-Butoxyethanol 99%
2-Butoxyethanol

Used in industry for degreasing and detergent applications.


Purity: 99% CAS No.: 111-76-2 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 64°(147°F) Physical Form: Clear liquid Density: 0.902 Refractive Index: 1.4190 Appearance: Colorless Odor: Mild Melting Point: -79° Boiling Point: 171-173° Beilstein Reference: 1732511 Merck Reference: 14,1559 UN No.: 2810 MDL No.: MFCD00002884


Used in industry for degreasing and detergent applications.

Formula: CH₃(CH₂)₃OCH₂CH₂OH
MW: 118.18 g/mol
Boiling Pt: 171…173 °C
Melting Pt: –79 °C
Density: 0.902
Flash Pt: 64 °C(147 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002884
CAS Number: 111-76-2
EINECS: 203-905-0
UN: 2810
ADR: 6.1,III
Merck Index: 14,1559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a9d2c527d0c167d2248d1936e2d1bd4aeab7a&vid=75260b2608ddc5eb29642178ab193fbcaf54198d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR