Dioctyl sulfide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dioctylsulfane, Dioctylsulphane

A17955-14 A17955-22 A17955-36
CAAAA17955-14EA 69.68 CAD
CAAAA17955-14 CAAAA17955-22 CAAAA17955-36
Dioctyl sulfide 97%
Dioctyl sulphide

Purity: 97% CAS No.: 2690-08-6 Molecular Formula: [CH3(CH2)7]2S Formula Weight: 258.51 Flash Point: 160°(320°F) Refractive Index: 1.4624 Odor: Stench Melting Point: 0-1° Boiling Point: 179-180°/10mm Beilstein Reference: 1701624 MDL No.: MFCD00009566

Formula: C₁₆H₃₄S
MW: 258.51 g/mol
Boiling Pt: 179…180 °C/10 mm
Melting Pt: 0…1 °C
Density: 0.844
Flash Pt: 160 °C(320 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009566
CAS Number: 2690-08-6
EINECS: 220-259-5
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR