Tris(3-methoxyphenyl)phosphine 98%

Supplier: Thermo Scientific Chemicals
Warning

A17949-03 A17949-06
CAAAA17949-03EA 113.44 CAD
CAAAA17949-03 CAAAA17949-06
Tris(3-methoxyphenyl)phosphine 98%
Tris(3-methoxyphenyl)phosphine

Purity: 98% CAS No.: 29949-84-6 Molecular Formula: C21H21O3P Formula Weight: 352.37 Melting Point: 113-115° MDL No.: MFCD00008386

Formula: C₂₁H₂₁O₃P
MW: 352.37 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008386
CAS Number: 29949-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR