Dithizone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17947-14
A17947-18
CAAAAA17947-06EA
52.88
CAD
CAAAAA17947-06
CAAAAA17947-14
CAAAA17947-18
Dithizone 98%
Dithizone
Purity: 98%
CAS No.: 60-10-6
Molecular Formula: C13H12N4S
Formula Weight: 256.33
Odor: Odorless
Melting Point: ca 168° dec.
Beilstein Reference: 748838
Merck Reference: 14,3377
MDL No.: MFCD00003025
Formula:
C₆H₅NHNHCSN=NC₆H₅ Boiling Pt: 406 °C (1013 hPa) Melting Pt: ∼ 168 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003025 CAS Number: 60-10-6 EINECS: 200-454-1 Merck Index: 14,3377 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...