4-Fluoroveratrole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dimethoxy-4-fluorobenzene, 4-Fluoro-1,2-dimethoxybenzene

A17936-06 A17936-14
CAAAA17936-06EA 59.41 CAD
CAAAA17936-06 CAAAA17936-14
4-Fluoroveratrole 98%
4-Fluoroveratrole

Purity: 98% CAS No.: 398-62-9 Molecular Formula: C8H9FO2 Formula Weight: 156.16 Flash Point: 91°(195°F) Refractive Index: 1.5100 Boiling Point: 120-123°/45mm Beilstein Reference: 2441600 MDL No.: MFCD00012201

Formula: C₈H₉FO₂
Boiling Pt: 120…123 °C (45 mmHg)
Density: 1,171
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012201
CAS Number: 398-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR