Polyethylene glycol 20,000
Supplier: Thermo Scientific Chemicals
A17925-0B
A17925-30
CAAAA17925-0BEA
243.32
CAD
CAAAA17925-0B
CAAAA17925-30
Polyethylene glycol 20,000
Polyethylene glycol 20,000
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 260°(500°F)
Physical Form: Powder or flake
Density: 1.20
Appearance: White
Odor: Odorless
pH: 5-7
Melting Point: 63-66°
Merck Reference: 14,7568
MDL No.: MFCD00081839
Formula:
H(OCH₂CH₂)nOH Melting Pt: 63…66 °C Density: 1.20 Flash Pt: 260 °C(500 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 203-473-3 Merck Index: 14,7568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...