Dodecyltrimethylammonium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC

A17924-22 A17924-06 A17924-14
CAAAAA17924-22EA 848.46 CAD
CAAAAA17924-22 CAAAA17924-06 CAAAA17924-14
Dodecyltrimethylammonium chloride 97%
Dodecyltrimethylammonium chloride

Purity: 97% CAS No.: 112-00-5 Molecular Formula: C15H34ClN Formula Weight: 263.90 Odor: Odorless Melting Point: ca 237° dec. Beilstein Reference: 3915951 UN No.: 3077 MDL No.: MFCD00041974

Formula: C₁₅H₃₄ClN
MW: 263.89 g/mol
Melting Pt: ∼ 237 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00041974
CAS Number: 112-00-5
EINECS: 203-927-0
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR