Heptylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17922-18
A17922-30
A17922-14
CAAAAA17922-18EA
89.43
CAD
CAAAAA17922-18
CAAAAA17922-30
CAAAA17922-14
Heptylamine 98+%
Heptylamine
Purity: 98+%
CAS No.: 111-68-2
Molecular Formula: C7H17N
Formula Weight: 115.22
Flash Point: 35°(95°F)
Refractive Index: 1.4245
Odor: Amine-like
Melting Point: -23°
Boiling Point: 154-156°
Beilstein Reference: 1731688
UN No.: 2734
MDL No.: MFCD00008244
Formula:
H₃C(CH₂)₅CH₂NH₂ Boiling Pt: 154…156 °C Melting Pt: –23 °C Density: 0.775 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008244 CAS Number: 111-68-2 EINECS: 203-895-8 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...