2,4-Difluorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17920-06
A17920-14
CAAAA17920-06EA
56.25
CAD
CAAAA17920-06
CAAAA17920-14
2,4-Difluorobenzoyl chloride 98%
2,4-Difluorobenzoyl chloride
Purity: 98%
CAS No.: 72482-64-5
Molecular Formula: C7H3ClF2O
Formula Weight: 176.55
Flash Point: 82°(179°F)
Refractive Index: 1.5145
Boiling Point: 96-98°/15mm
Beilstein Reference: 4800566
UN No.: 3265
MDL No.: MFCD00000658
Formula:
C₇H₃ClF₂O MW: 176.55 Boiling Pt: 68 Density: 1.437 Flash Pt: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000658 CAS Number: 72482-64-5 EINECS: 276-690-4 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...