Bromothymol blue sodium salt

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulfonate, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate

A17913-09 A17913-18
CAAAA17913-09EA 75.37 CAD
CAAAA17913-09 CAAAA17913-18
Bromothymol blue sodium salt
Bromothymol blue sodium salt

CAS No.: 34722-90-2 Molecular Formula: C27H27Br2NaO5S Formula Weight: 646.38 Melting Point: 200-202° MDL No.: MFCD00077263

Formula: C₂₇H₂₇Br₂NaO₅S
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR