1-Ethylpyridinium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethylpyridin-1-ium bromide

A17911-14 A17911-22
CAAAA17911-14EA 103.02 CAD
CAAAA17911-14 CAAAA17911-22
1-Ethylpyridinium bromide 99%
1-Ethylpyridinium bromide

Purity: 99% CAS No.: 1906-79-2 Molecular Formula: C7H10BrN Formula Weight: 188.07 Melting Point: 117-121° MDL No.: MFCD00041999

Formula: C₇H₁₀BrN
MW: 188.07 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00041999
CAS Number: 1906-79-2
EINECS: 217-607-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR