Wright's stain for use in preparing solutions of leishman's, jenner's, giemsa's and other stains

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

A17903-14 A17903-22
CAAAA17903-14EA 105.39 CAD
CAAAA17903-14 CAAAA17903-22
Wright's stain for use in preparing solutions of leishman's, jenner's, giemsa's and other stains
Wright's stain

Useful stain for blood and for bone marrow films


CAS No.: 68988-92-1 MDL No.: MFCD00082143

Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
EINECS: 273-541-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR